missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metaldehyde, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Metaldehyde |
| CAS | 108-62-3 |
| Molecular Formula | C8 H16 O4 |
| Quantity | 1 g |
| Synonym | Metaldehyde, 2,4,6,8-Tetramethyl-1,3,5,7-tetroxocane, Acetaldehyde Tetramer |
| InChI Formula | InChI=1S/C8H16O4/c1-5-9-6(2)11-8(4)12-7(3)10-5/h5-8H,1-4H3 |
| SMILES | CC1OC(C)OC(C)OC(C)O1 |
| IUPAC Name | 2,4,6,8-tetramethyl-1,3,5,7-tetraoxocane |
| Molecular Weight (g/mol) | 176.2102 |
| Formula Weight | 176.1049 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction