missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metals in Biodiesel Standard: Ca, K, Mg, Na, P @ 20 μg/g in B100 Biodiesel, VHG™
Description
Metals in Biodiesel Standard: Ca, K, Mg, Na, P @ 20 μg/g in B100 Biodiesel
Specifications
Specifications
| Chemical Name or Material | Calcium/Magnesium/Phosphorus/Potassium/Sodium |
| CAS | 7440-70-2, 7439-95-4, 7723-14-0, 9-7-7440, 7440-23-5 |
| Analyte or Component Names | Calcium/Magnesium/Phosphorus/Potassium/Sodium |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | B100 Biodiesel |
| Quantity | 100 g |
| Concentration or Composition (by Analyte or Components) | Calcium 20 μg/g/Magnesium 20 μg/g/Phosphorus 20 μg/g/Potassium 20 μg/g/Sodium 20 μg/g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?