missing translation for 'onlineSavingsMsg'
Learn More

Methanol, CHROMASOLV™, gradient grade, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™

Product Code. 15674680
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
Unit Size:
2.5 L
Pack of 4
This item is not returnable. View return policy

Product Code. 15674680

Brand: Honeywell-Chromasolv 34885E4X2.5L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % ≤100.0000%
Quantity 4 x 2.5 L
Packaging Glass Bottle
UN Number UN1230
Beilstein 1098229
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Methanol, CHROMASOLV, gradient grade, for HPLC, 99.9%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.