missing translation for 'onlineSavingsMsg'
Learn More

Methanol, LC-MS Grade, 99.8+%, Thermo Scientific™

Solvent for LC-MS analysis

60.65€ - 195.65€

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15492897
SDS View Documents
Thermo Scientific Alfa Aesar
047192.K2
1L
ONLINE EXCLUSIVE PRICE
67.20€
60.65€ / 1 L
Save 6.55€
9% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15412907
SDS View Documents
Thermo Scientific Alfa Aesar
047192.M1
4 x 1L
ONLINE EXCLUSIVE PRICE
217.00€
195.65€ / 1 L
Save 21.35€
9% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

67-56-1
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO

Specifications

67-56-1
11°C (52°F)
1.329
CH3OH
UN1230
14,5957
Miscible with water and most of the organic solvents including ethyl acetate,toluene,dichloromethane,chloroform,tetrahydrofuran,and acetone.
CO
32.04
CHEBI:17790
≥99.8%
Methanol
0.791 g/mL
Mild
CH4O
MFCD00004595
1098229
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
LC-MS
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H301-H311-H331-H370
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c

H225-H301+H311+H331-H370-H372

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL

missing translation for 'einecsNumber' : 200-659-6

missing translation for 'rtecsNumber' : PC1400000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Methanol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

Supplemental information
  • MIXTURE LIST-Contain: methanol

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.