missing translation for 'onlineSavingsMsg'
Learn More

Methazolamide, TRC

Product Code. 30136440
Change view
Click to view available options
Quantity:
50 mg
1 g
2.5 g
Unit Size:
1g
2.50g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136440 2.5 g 2.50g
30005615 50 mg 50mg
30006428 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136440 Supplier Toronto Research Chemicals Supplier No. TRCM2260202.5G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-57-4
Molecular Formula C5 H8 N4 O3 S2
Molecular Weight (g/mol) 236.27
Synonym Acetamide, N-(4-methyl-2-sulfamoyl-δ2-1,3,4-thiadiazolin-5-ylidene)- (6CI,7CI,8CI), N-[5-(Aminosulfonyl)-3-methyl-1,3,4-thiadiazol-2(3H)-ylidene]acetamide, 5-Acetylimino-4-methyl-δ2-1,3,4-thiadiazoline-2-sulfonamide, L 584601, Methazolamide, Methenamide, N-(4-Methyl-2-sulfamoyl-δ2-1,3,4-thiadiazolin-5-ylidene)acetamide, Neptazane, Neptazaneat
IUPAC Name (NZ)-N-(3-methyl-5-sulfamoyl-1,3,4-thiadiazol-2-ylidene)acetamide
SMILES CN1N=C(S/C/1=N\C(=O)C)S(=O)(=O)N

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C5H8N4O3S2/c1-3(10)7-4-9(2)8-5(13-4)14(6,11)12/h1-2H3,(H2,6,11,12)/b7-4-
Formula Weight 236.0038
Chemical Name or Material Methazolamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.