missing translation for 'onlineSavingsMsg'
Learn More

Methiocarb, TRC

Product Code. 30138385
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138385 10 g 10g
30040515 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138385 Supplier Toronto Research Chemicals Supplier No. TRCM22524010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2032-65-7
Molecular Formula C11 H15 N O2 S
Molecular Weight (g/mol) 225.31
Synonym Carbamic acid, methyl-, 4-(methylthio)-3,5-xylyl ester (6CI,7CI,8CI), Phenol, 3,5-dimethyl-4-(methylthio)-, methylcarbamate (9CI), 3,5-Dimethyl-4-(methylthio)phenyl methylcarbamate, 4-(Methylthio)-3,5-xylyl methylcarbamate, 4-(Methylthio)-3,5-xylyl-N-methylcarbamate, 4-Methylthio-3,5-dimethylphenyl methylcarbamate, B 37344, BAY 37344, BAY 5024, DCR 736, Draza, Mercaptodimethur, Mesurol, Mesurol 50, Mesurol 500FS, Mesurol phenol, Methiocarb, Metmercapturon, SD 9228, Phenol, 3,5-dimethyl-4-(methylthio)-, 1-(N-methylcarbamate)
IUPAC Name (3,5-dimethyl-4-methylsulfanylphenyl) N-methylcarbamate
SMILES CNC(=O)Oc1cc(C)c(SC)c(C)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H15NO2S/c1-7-5-9(14-11(13)12-3)6-8(2)10(7)15-4/h5-6H,1-4H3,(H,12,13)
Formula Weight 225.0823
Chemical Name or Material Methiocarb
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.