missing translation for 'onlineSavingsMsg'
Learn More

Methomyl, TRC

Product Code. 30136066
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136066 50 g 50g
30136901 1 g 1g
17967169 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136066 Supplier Toronto Research Chemicals Supplier No. TRCM26053050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16752-77-5
Molecular Formula C5 H10 N2 O2 S
Molecular Weight (g/mol) 162.21
Synonym Ethanimidothioic acid, N-[[(methylamino)carbonyl]oxy]-, methyl ester, Du Pont 1179, Du Pont Insecticide 1179, Dunet, IN 1179, Lan Bait, Lannate, Lannate 20, Lannate 20L, Lannate 25, Lannate 90SP, Lannate D, Lannate L, Lannate LV, Mesomile, Methomyl, Methomyl SP, Methyl N-[(methylcarbamoyl)oxy]thioacetimidate, Methyl O-(methylcarbamoyl)thiolacetohydroxamate, Methyl O-(methylcarbamyl)thiolacetohydroxamate, Metomil, Metomur, Mie Duo Wei, Nudrin, Nudrin 225, Nudrin D, S-Methyl N-[(methylcarbamoyl)oxy]thioacetimidate, SD 14999, WL 18236
IUPAC Name methyl N-(methylcarbamoyloxy)ethanimidothioate
SMILES CNC(=O)ON=C(C)SC

Specifications

Quantity 50 g
InChI Formula InChI=1S/C5H10N2O2S/c1-4(10-3)7-9-5(8)6-2/h1-3H3,(H,6,8)
Formula Weight 162.0463
Percent Purity >95%
Chemical Name or Material Methomyl
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.