Learn More
Methyl 1-(2,4-Difluorophenyl)-3-(thiophen-2-yl)-1H-pyrazole-5-carboxylate, TRC
Product Code. 30478955
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478955 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30478955 Supplier Toronto Research Chemicals Supplier No. TRCM33423310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl 1-(2,4-Difluorophenyl)-3-(thiophen-2-yl)-1H-pyrazole-5-carboxylate
CAS 1202028-96-3
Molecular Formula C15 H10 F2 N2 O2 S
Quantity 10 mg
InChI Formula InChI=1S/C15H10F2N2O2S/c1-21-15(20)13-8-11(14-3-2-6-22-14)18-19(13)12-5-4-9(16)7-10(12)17/h2-8H,1H3
SMILES COC(=O)c1cc(nn1c2ccc(F)cc2F)c3cccs3
IUPAC Name methyl 2-(2,4-difluorophenyl)-5-thiophen-2-ylpyrazole-3-carboxylate
Molecular Weight (g/mol) 320.314
Formula Weight 320.043
Product Title
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.