missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-methylpyrrole-2-carboxylate, 99%

Product Code. 11474424
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474424 25 g 25g
11464424 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474424 Supplier Thermo Scientific Alfa Aesar Supplier No. A13095.14
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 37619-24-2 | C7H9NO2 | 139.154 g/mol

Methyl 1-methylpyrrole-2-carboxylate is used as chemical and organic Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 1-methylpyrrole-2-carboxylate is used as chemical and organic Intermediates.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37619-24-2
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD00052747
InChI Key APHVGKYWHWFAQV-UHFFFAOYSA-N
Synonym methyl 1-methyl-1h-pyrrole-2-carboxylate, methyl1-methyl-1h-pyrrole-2-carboxylate, 1-methyl-1h-pyrrole-2-carboxylic acid methyl ester, methyl 1-methyl-2-pyrrolecarboxylate, 1-methylpyrrole-2-carboxylic acid methyl ester, pubchem12438, acmc-209iue, methylmethylpyrrolecarboxylate, 2-methoxycarbonyl-1-methylpyrrole, methyl 1-methyl-pyrrole-2-carboxylate
PubChem CID 142178
IUPAC Name methyl 1-methylpyrrole-2-carboxylate
SMILES CN1C=CC=C1C(=O)OC

Specifications

Boiling Point 96°C to 98°C (15 mmHg)
Refractive Index 1.522
Quantity 25 g
Beilstein 116477
Solubility Information Slightly soluble in water.
Formula Weight 139.16
Percent Purity 99%
Chemical Name or Material Methyl 1-methylpyrrole-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.