Learn More
Methyl 2-[3-Cyano-4-(2-methylpropoxy)phenyl]-4-methylthiazole-5-carboxylate, TRC
Product Code. 30137656
Change view
Click to view available options
Quantity:
10 mg
500 mg
1 g
Unit Size:
10mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137656 500 mg 500mg
30136588 1 g 1g
30005910 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137656 Supplier Toronto Research Chemicals Supplier No. TRCM296485500MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 923942-34-1
Molecular Formula C17H18N2O3S
Molecular Weight (g/mol) 330.4
Synonym Methyl 2-[3-Cyano-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid Ester, Methyl 2-(3-Cyano-4-isobutoxyphenyl)-4-methylthiazole-5-carboxylate Acetate;
IUPAC Name methyl 2-[3-cyano-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carboxylate
SMILES CC1=C(C(OC)=O)SC(C2=CC(C#N)=C(OCC(C)C)C=C2)=N1
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C17H18N2O3S/c1-10(2)9-22-14-6-5-12(7-13(14)8-18)16-19-11(3)15(23-16)17(20)21-4/h5-7,10H,9H2,1-4H3
Formula Weight 330.1038
Chemical Name or Material Methyl 2-[3-Cyano-4-(2-methylpropoxy)phenyl]-4-methylthiazole-5-carboxylate