Learn More
Methyl 2,3-O-Isopropylidene-Beta-D-ribofuranoside, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Methyl 2,3-O-Isopropylidene-beta-D-ribofuranoside |
| CAS | 4099-85-8 |
| Molecular Formula | C9 H16 O5 |
| Quantity | 5 g |
| Synonym | Capecitabine Impurity 32 |
| InChI Formula | InChI=1S/C9H16O5/c1-9(2)13-6-5(4-10)12-8(11-3)7(6)14-9/h5-8,10H,4H2,1-3H3/t5-,6-,7-,8-/m1/s1 |
| SMILES | CO[C@@H]1O[C@H](CO)[C@H]2OC(C)(C)O[C@@H]12 |
| IUPAC Name | [(3aR,4R,6R,6aR)-4-methoxy-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-6-yl]methanol |
| Molecular Weight (g/mol) | 204.22 |
| Formula Weight | 204.0998 |