missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Chloroacetoacetate, TRC

Product Code. 30467841
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30467841 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30467841 Supplier Toronto Research Chemicals Supplier No. TRCM2955625G
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 2-Chloroacetoacetate
CAS 4755-81-1
Molecular Formula C5H7ClO3
Quantity 5 g
Synonym 2-Chloro-3-oxo-butyric Acid Methyl Ester, 2-Chloro-3-oxobutanoic Acid Methyl Ester, 2-Chloroacetoacetic Acid Methyl Ester, Methyl 2-Chloro-3-oxobutanoate, Methyl 2-Chloro-3-oxobutyrate, Methyl 2-Chloroacetoacetate, Methyl alpha-Chloroacetoacetate
InChI Formula InChI=1S/C5H7ClO3/c1-3(7)4(6)5(8)9-2/h4H,1-2H3
SMILES COC(=O)C(Cl)C(=O)C
IUPAC Name methyl 2-chloro-3-oxobutanoate
Molecular Weight (g/mol) 150.56
Formula Weight 150.0084
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.