missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-fluorobenzoate, 98%, BLD Pharm™

Product Code. 30988470
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
1 kg
Unit Size:
1kg
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30988470 10 g -
30981033 100 g -
30982757 500 g -
30980270 25 g -
30973986 5 g 5g
30987240 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30988470 Supplier BLD Pharm Supplier No. BD13828310g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 394-35-4
Molecular Formula C8H7FO2
MDL Number MFCD00017913
Quantity 10 g
SMILES C1=CC=CC(=C1F)C(OC)=O
Molecular Weight (g/mol) 154.14
Percent Purity 98%
Chemical Name or Material Methyl 2-fluorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.