missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Nitro-4-(trifluoromethyl)benzoate, TRC

Product Code. 30180018
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180018 25 g 25g
30179724 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180018 Supplier Toronto Research Chemicals Supplier No. TRCM32448025G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 228418-45-9
Molecular Formula C9H6F3NO4
Molecular Weight (g/mol) 249.15
Synonym 2-Nitro-4-trifluoromethylbenzoic Acid Methyl Ester, Methyl 2-Nitro-4-trifluoromethylbenzoate
IUPAC Name methyl 2-nitro-4-(trifluoromethyl)benzoate
SMILES COC(=O)c1ccc(cc1[N+](=O)[O-])C(F)(F)F

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H6F3NO4/c1-17-8(14)6-3-2-5(9(10,11)12)4-7(6)13(15)16/h2-4H,1H3
Formula Weight 249.0249
Chemical Name or Material Methyl 2-Nitro-4-(trifluoromethyl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.