missing translation for 'onlineSavingsMsg'
Learn More

Methyl (2S)-2-Aminobutanoate, TRC

Product Code. 30472664
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30472664 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30472664 Supplier Toronto Research Chemicals Supplier No. TRCM285735500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl (2S)-2-Aminobutanoate
CAS 15399-22-1
Molecular Formula C5 H11 N O2
Quantity 500 mg
Synonym Butanoic acid','2-amino-','methyl ester','(2S)-, Butanoic acid','2-amino-','methyl ester','(S)-, Butyric acid','2-amino-','methyl ester','L- (8CI), Methyl (2S)-2-aminobutanoate, (S)-2-Aminobutanoic acid methyl ester, (S)-Methyl 2-aminobutanoate, L-2-Aminobutyric a
InChI Formula InChI=1S/C5H11NO2/c1-3-4(6)5(7)8-2/h4H,3,6H2,1-2H3/t4-/m0/s1
SMILES CC[C@H](N)C(=O)OC
IUPAC Name methyl (2S)-2-aminobutanoate
Molecular Weight (g/mol) 117.146
Formula Weight 117.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.