missing translation for 'onlineSavingsMsg'
Learn More

Methyl (2S,4S)-1-Boc-4-aminopyrrolidine-2-carboxylate hydrochloride, 97%, BLD Pharm™

Product Code. 30801626
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
25 g
Unit Size:
100mg
1g
250mg
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30801626 1 g 1g
30791419 25 g 25g
30797193 250 mg 250mg
30798127 5 g 5g
30792317 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30801626 Supplier BLD Pharm Supplier No. BD1436631g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 171110-72-8
Molecular Formula C11H21ClN2O4
MDL Number MFCD03787927
Quantity 1 g
SMILES O=C([C@H]1N(C(OC(C)(C)C)=O)C[C@@H](N)C1)OC.[H]Cl
Molecular Weight (g/mol) 280.75
Percent Purity 97%
Chemical Name or Material Methyl (2S,4S)-1-Boc-4-aminopyrrolidine-2-carboxylate hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.