missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(3’,4’-Dimethoxyphenyl)propanoate, TRC

Product Code. 30467007
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30467007 100 mg 100mg
30478635 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30467007 Supplier Toronto Research Chemicals Supplier No. TRCM302865100MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27798-73-8
Molecular Formula C12 H16 O4
Molecular Weight (g/mol) 224.25
IUPAC Name methyl 3-(3,4-dimethoxyphenyl)propanoate
SMILES COC(=O)CCc1ccc(OC)c(OC)c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H16O4/c1-14-10-6-4-9(8-11(10)15-2)5-7-12(13)16-3/h4,6,8H,5,7H2,1-3H3
Formula Weight 224.1049
Chemical Name or Material Methyl 3-(3',4'-Dimethoxyphenyl)propanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.