Learn More
Methyl 3-(3’,4’-Dimethoxyphenyl)propanoate, TRC
Product Code. 30478635
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478635 500 mg 500mg
30467007 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30478635 Supplier Toronto Research Chemicals Supplier No. TRCM302865500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27798-73-8
Molecular Formula C12 H16 O4
Molecular Weight (g/mol) 224.25
IUPAC Name methyl 3-(3,4-dimethoxyphenyl)propanoate
SMILES COC(=O)CCc1ccc(OC)c(OC)c1
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C12H16O4/c1-14-10-6-4-9(8-11(10)15-2)5-7-12(13)16-3/h4,6,8H,5,7H2,1-3H3
Formula Weight 224.1049
Chemical Name or Material Methyl 3-(3',4'-Dimethoxyphenyl)propanoate