missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-difluorobenzoate, 98%

Product Code. 11343986
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343986 5 g 5g
11353986 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343986 Supplier Thermo Scientific Alfa Aesar Supplier No. L17171.06
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 216393-55-4 | C8H6F2O2 | 172.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216393-55-4
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.13
MDL Number MFCD01318160
InChI Key JBEJPGWPXIIQBB-UHFFFAOYSA-N
PubChem CID 2775360
IUPAC Name methyl 3,5-difluorobenzoate
SMILES COC(=O)C1=CC(F)=CC(F)=C1

Specifications

Refractive Index 1.474
Quantity 5 g
Synonym 3,5-difluorobenzoic acid methyl ester, benzoic acid, 3,5-difluoro-, methyl ester, acmc-209fmq, methyl-3,5-difluorophenylacetate, methyl-3,5-difluorobenzoate, 3,5-difluorobenzoicacid methyl ester, benzoic acid,3,5-difluoro-, methyl ester
Formula Weight 172.13
Percent Purity 98%
Chemical Name or Material Methyl 3,5-difluorobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.