Learn More
Methyl 3-Hydroxyphenylacetate, TRC
Product Code. 30010013
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010013 10 g 10g
30011463 1 g 1g
30011490 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010013 Supplier Toronto Research Chemicals Supplier No. TRCM31278510G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 42058-59-3
Molecular Formula C26 H27 Cl O10
Molecular Weight (g/mol) 534.94
Synonym 3-Hydroxybenzeneacetic Acid Methyl Ester, (3-Hydroxyphenyl)acetic Acid Methyl ester, 2-(3-Hydroxyphenyl)acetic Acid Methyl Ester, Methyl 2-(3-Hydroxyphenyl)acetate, Methyl 3-Hydroxybenzeneacetate, Methyl m-hydroxyphenylacetate
IUPAC Name prop-2-enyl (3R,6S)-6-[2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoyl]oxy-3,4,5-trihydroxyoxane-2-carboxylate
SMILES CC(C)(Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2)C(=O)O[C@@H]3OC([C@H](O)C(O)C3O)C(=O)OCC=C
Specifications
Quantity 10 g
InChI Formula InChI=1S/C26H27ClO10/c1-4-13-34-23(32)22-20(30)19(29)21(31)24(35-22)36-25(33)26(2,3)37-17-11-7-15(8-12-17)18(28)14-5-9-16(27)10-6-14/h4-12,19-22,24,29-31H,1,13H2,2-3H3/t19?,20-,21?,22?,24+/m1/s1
Formula Weight 534.129
Chemical Name or Material Methyl 3-Hydroxyphenylacetate