missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Hydroxyphenylacetate, TRC

Product Code. 30011490
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011490 5 g 5g
30010013 10 g 10g
30011463 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011490 Supplier Toronto Research Chemicals Supplier No. TRCM3127855G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42058-59-3
Molecular Formula C26 H27 Cl O10
Molecular Weight (g/mol) 534.94
Synonym 3-Hydroxybenzeneacetic Acid Methyl Ester, (3-Hydroxyphenyl)acetic Acid Methyl ester, 2-(3-Hydroxyphenyl)acetic Acid Methyl Ester, Methyl 2-(3-Hydroxyphenyl)acetate, Methyl 3-Hydroxybenzeneacetate, Methyl m-hydroxyphenylacetate
IUPAC Name prop-2-enyl (3R,6S)-6-[2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoyl]oxy-3,4,5-trihydroxyoxane-2-carboxylate
SMILES CC(C)(Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2)C(=O)O[C@@H]3OC([C@H](O)C(O)C3O)C(=O)OCC=C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C26H27ClO10/c1-4-13-34-23(32)22-20(30)19(29)21(31)24(35-22)36-25(33)26(2,3)37-17-11-7-15(8-12-17)18(28)14-5-9-16(27)10-6-14/h4-12,19-22,24,29-31H,1,13H2,2-3H3/t19?,20-,21?,22?,24+/m1/s1
Formula Weight 534.129
Chemical Name or Material Methyl 3-Hydroxyphenylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.