missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Phenylpropionate(3-Phenylpropionic Acid Methyl Ester), TRC

Product Code. 30428753
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30428753 10 g
30422615 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30428753 Supplier Toronto Research Chemicals Supplier No. TRCP33619010G
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-25-3
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
Synonym Benzenepropanoic Acid Methyl Ester, 3-Phenylpropanoic Acid Methyl Ester, 3-Phenylpropionic Acid Methyl Ester, Dihydrocinnamic Acid Methyl Ester, Methyl 3-phenylpropanoate, Methyl benzenepropanoate, Methyl dihydrocinnamate, Methyl Hydrocinnamate, Methyl beta-Phenylpropionate, NSC 10128, NSC 404188, beta-Phenylpropionic Acid Methyl Ester
IUPAC Name methyl 3-phenylpropanoate
SMILES COC(=O)CCc1ccccc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H12O2/c1-12-10(11)8-7-9-5-3-2-4-6-9/h2-6H,7-8H2,1H3
Formula Weight 164.0837
Chemical Name or Material Methyl 3-Phenylpropionate(3-Phenylpropionic Acid Methyl Ester)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.