missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(trimethylsilyl)propiolate, 98%, BLD Pharm™

Product Code. 30985318
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30985318 250 mg -
30981302 5 g -
30972434 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30985318 Supplier BLD Pharm Supplier No. BD138470250mg
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 42201-71-8
Molecular Formula C7H12O2Si
MDL Number MFCD04039783
Quantity 250 mg
SMILES O=C(OC)C#C[Si](C)(C)C
Molecular Weight (g/mol) 156.25
Percent Purity 98%
Chemical Name or Material Methyl 3-(trimethylsilyl)propiolate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.