missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-[(5-Amino-1-methylindol-3-yl)methyl]-3-methoxybenzoate, TRC

Product Code. 30007001
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007001 100 mg 100mg
30009673 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007001 Supplier Toronto Research Chemicals Supplier No. TRCM258650100MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107754-14-3
Molecular Formula C19 H20 N2 O3
Molecular Weight (g/mol) 324.37
Synonym 4-[(5-Amino-1-methyl-1H-indol-3-yl)methyl]-3-methoxy-benzoic Acid Methyl Ester
IUPAC Name methyl 4-[(5-amino-1-methyl-indol-3-yl)methyl]-3-methoxy-benzoate
SMILES COC(=O)c1ccc(Cc2cn(C)c3ccc(N)cc23)c(OC)c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H20N2O3/c1-21-11-14(16-10-15(20)6-7-17(16)21)8-12-4-5-13(19(22)24-3)9-18(12)23-2/h4-7,9-11H,8,20H2,1-3H3
Formula Weight 324.1474
Chemical Name or Material Methyl 4-[(5-Amino-1-methylindol-3-yl)methyl]-3-methoxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.