Learn More
Methyl 4-Methoxypicolinate, TRC
Product Code. 30483508
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483508 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30483508 Supplier Toronto Research Chemicals Supplier No. TRCM3384941G
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl 4-Methoxypicolinate
CAS 29681-43-4
Molecular Formula C8H9NO3
Quantity 1 g
Synonym 4-Methoxypyridine-2-carboxylic Acid Methyl Ester, Methyl 4-(Methyloxy)-2-pyridinecarboxylate, Methyl 4-Methoxy-2-picolinate, Methyl 4-Methoxypicolinate, Methyl 4-Methoxypyridine-2-carboxylate
InChI Formula InChI=1S/C8H9NO3/c1-11-6-3-4-9-7(5-6)8(10)12-2/h3-5H,1-2H3
SMILES COC(=O)c1cc(OC)ccn1
IUPAC Name methyl 4-methoxypyridine-2-carboxylate
Molecular Weight (g/mol) 167.16
Formula Weight 167.0582