missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-tert-butylbenzoate, 99%, Thermo Scientific™

Product Code. 10684662 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 10684662

Brand: Thermo Scientific Acros 173480250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 26537-19-9
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 122.0°C to 124.0°C (9.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C12H16O2
Linear Formula (CH3)3CC6H4CO2CH3
Refractive Index 1.5090 to 1.5110
MDL Number MFCD00008835
Quantity 25 g
Beilstein 09, 560
Specific Gravity 0.99
Synonym methyl 4-tert-butyl benzoate, methyl p-tert-butylbenzoate, 4-tert-butylbenzoic acid methyl ester, unii-8vk4o27jmt, benzoic acid, 4-1,1-dimethylethyl-, methyl ester, methyl 4-t-butylbenzoate, 8vk4o27jmt, methyl4-tert-butylbenzoate, methyl p-t-butylbenzoate, benzoic acid, p-tert-butyl-, methyl ester
InChI Key UPIJOAFHOIWPLT-UHFFFAOYSA-N
SMILES CC(C)(C)C1=CC=C(C=C1)C(=O)OC
IUPAC Name methyl 4-tert-butylbenzoate
Molecular Weight (g/mol) 192.26
PubChem CID 97433
Formula Weight 192.26
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl 4-tert-butylbenzoate, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.