Learn More
Methyl 5-acetyl-2-hydroxybenzoate, 98%, BLD Pharm™
Product Code. 30783395
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Quantity:
5 g
10 g
25 g
100 g
500 g
Förpackningsstorlek:
100g
10g
25g
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30783395 10 g 10g
30772051 5 g 5g
30766898 100 g 100g
30781782 500 g 500g
30768834 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30783395 Supplier BLD Pharm Supplier No. BD1185310g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
CAS 16475-90-4
Molecular Formula C10H10O4
MDL Number MFCD00040924
Quantity 10 g
SMILES COC(=O)C1=CC(=CC=C1O)C(C)=O
Molecular Weight (g/mol) 194.18
Percent Purity 98%
Chemical Name or Material Methyl 5-acetyl-2-hydroxybenzoate