missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-bromo-2-naphthoate, 98%

Product Code. 10642803 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10642803 25 g Glass bottle 25g
10405292 5 g Glass bottle 5g
11974991 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10642803 Supplier Thermo Scientific Acros Supplier No. 364140250
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 33626-98-1 | C12H9BrO2 | 265.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33626-98-1
Molecular Formula C12H9BrO2
Molecular Weight (g/mol) 265.1
MDL Number MFCD00100408
InChI Key JEUBRLPXJZOGPX-UHFFFAOYSA-N
PubChem CID 854134
IUPAC Name methyl 6-bromonaphthalene-2-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=C(C=C2)Br

Specifications

Melting Point 123.0°C to 126.0°C
Color Brown or Cream to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC10H6COOCH3
Quantity 25 g
Synonym methyl 6-bromo-2-naphthoate, methyl6-bromo-2-naphthoate, 6-bromo-2-naphthalenecarboxylic acid methyl ester, 6-bromo-2-naphthoic acid methyl ester, 2-naphthalenecarboxylic acid, 6-bromo-, methyl ester, 6-bromonaphthalene-2-carboxylic acid methyl ester, 6-bromo-naphthalene-2-carboxylic acid methyl ester, pubchem9467, acmc-209i1z, 6-bromo-2-methylnaphthoate
Formula Weight 265.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-bromo-2-naphthoate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.