missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-hydroxy-2,2-dimethyl-2H-benzo[h]chromene-5-carboxylate, 98%, BLD Pharm™

Product Code. 31006576
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31006576 100 mg 100mg
30989370 250 mg 250mg
30994753 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31006576 Supplier BLD Pharm Supplier No. BD140248100mg
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 55481-88-4
Molecular Formula C17H16O4
MDL Number MFCD02752331
Quantity 100 mg
SMILES O=C(C1=C(C=CC(C)(C)O2)C2=C3C=CC=CC3=C1O)OC
Molecular Weight (g/mol) 284.31
Percent Purity 98%
Chemical Name or Material Methyl 6-hydroxy-2,2-dimethyl-2H-benzo[h]chromene-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.