Learn More
Methyl (6-(phenylsulfinyl)-1H-benzo[d]imidazol-2-yl)carbamate, 99%, BLD Pharm™
Product Code. 31007312
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
25 g
100 g
500 g
Unit Size:
100g
100mg
1g
25g
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31007312 500 g 500g
31004907 100 mg 100mg
30998839 5 g 5g
30993831 100 g 100g
30997425 1 g 1g
30994757 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31007312 Supplier BLD Pharm Supplier No. BD146493500g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
CAS 53716-50-0
Molecular Formula C15H13N3O3S
MDL Number MFCD00801063
Quantity 500 g
SMILES O=C(OC)NC1=NC2=CC=C(S(C3=CC=CC=C3)=O)C=C2N1
Molecular Weight (g/mol) 315.35
Percent Purity 99%
Chemical Name or Material Methyl (6-(phenylsulfinyl)-1H-benzo[d]imidazol-2-yl)carbamate