missing translation for 'onlineSavingsMsg'
Learn More

Methyl a-Bromophenylacetate, TRC

Product Code. 30473729
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30473729 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473729 Supplier Toronto Research Chemicals Supplier No. TRCM294940100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl alpha-Bromophenylacetate
CAS 3042-81-7
Molecular Formula C9H9BrO2
Quantity 100 g
Synonym alpha-Bromobenzeneacetic Acid Methyl Ester, Bromophenylacetic Acid Methyl Ester, 2-Bromo-2-phenylacetic Acid Methyl Ester, Methyl 2-Bromo-2-phenylacetate, Methyl 2-Bromophenylacetate, Methyl DL-Bromophenylacetate, Methyl Bromophenylacetate, Methyl alpha-Bromo-alpha-phenylacetate, Methyl alpha-Bromobenzeneacetate, NSC 243708, alpha-Bromobenzeneacetic Acid Methyl Ester, alpha-Bromophenylacetic Acid Methyl Ester
InChI Formula InChI=1S/C9H9BrO2/c1-12-9(11)8(10)7-5-3-2-4-6-7/h2-6,8H,1H3
SMILES COC(=O)C(Br)c1ccccc1
IUPAC Name methyl 2-bromo-2-phenylacetate
Molecular Weight (g/mol) 229.07
Formula Weight 227.9786
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.