Learn More
Methyl benzenesulfonate, 98%
Product Code. 11420338
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420338 500 g 500g
11400338 25 g 25g
11410338 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11420338 Supplier Thermo Scientific Alfa Aesar Supplier No. B24515.36
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 80-18-2 | C7H8O3S | 172.20 g/mol
Methyl benzenesulfonate is a sulfonate ester which acts as a potential genotoxic impurity in drug substances. Further, it exerts genotoxic effects in bacterial and mammalian cell systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl benzenesulfonate is a sulfonate ester which acts as a potential genotoxic impurity in drug substances. Further, it exerts genotoxic effects in bacterial and mammalian cell systems.
Solubility
Miscible with chloroform, methanol, ethanol and ether. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 80-18-2
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.20
MDL Number MFCD00014737
InChI Key CZXGXYBOQYQXQD-UHFFFAOYSA-N
Synonym benzenesulfonic acid, methyl ester, methyl benzenesulphonate, methyl besylate, methylbenzenesulfonate, ccris 9156, benzenesulfonic acid methyl ester, methyl benzene sulfonate, methyl ester of benzenesulphonic acid, methylbenzene sulfonate, pubchem22730
PubChem CID 6630
IUPAC Name methyl benzenesulfonate
SMILES COS(=O)(=O)C1=CC=CC=C1
Specifications
Melting Point -4°C
Density 1.269
Boiling Point 280°C
Flash Point 143°C (289°F)
Refractive Index 1.5165
Quantity 500 g
UN Number UN3265
Beilstein 908448
Solubility Information Miscible with chloroform,methanol,ethanol and ether. Immiscible with water.
Formula Weight 172.2
Percent Purity 98%
Chemical Name or Material Methyl benzenesulfonate