missing translation for 'onlineSavingsMsg'
Learn More

Methyl butyrate, TRC

Product Code. 30135149
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135149 25 g 25g
30138636 100 g 100g
30008150 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135149 Supplier Toronto Research Chemicals Supplier No. TRCM29504525G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-42-7
Molecular Formula C5 H10 O2
Molecular Weight (g/mol) 102.13
Synonym Butyric acid, ester with MeOH (4CI), Butyric acid, methyl ester (6CI,8CI), 3-Methylpropanoic acid methyl ester, Methyl butanoate, Methyl butyrate, Methyl n-butyrate, NSC 9380
IUPAC Name methyl butanoate
SMILES CCCC(=O)OC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C5H10O2/c1-3-4-5(6)7-2/h3-4H2,1-2H3
Formula Weight 102.0681
Chemical Name or Material Butyric acid methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.