Learn More
Methyl butyrate, TRC
Product Code. 30138636
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138636 100 g 100g
30135149 25 g 25g
30008150 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138636 Supplier Toronto Research Chemicals Supplier No. TRCM295045100G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 623-42-7
Molecular Formula C5 H10 O2
Molecular Weight (g/mol) 102.13
Synonym Butyric acid, ester with MeOH (4CI), Butyric acid, methyl ester (6CI,8CI), 3-Methylpropanoic acid methyl ester, Methyl butanoate, Methyl butyrate, Methyl n-butyrate, NSC 9380
IUPAC Name methyl butanoate
SMILES CCCC(=O)OC
Specifications
Quantity 100 g
InChI Formula InChI=1S/C5H10O2/c1-3-4-5(6)7-2/h3-4H2,1-2H3
Formula Weight 102.0681
Chemical Name or Material Butyric acid methyl ester