missing translation for 'onlineSavingsMsg'
Learn More

Methyl D-(+)-Lactate, TRC

Product Code. 30467468
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30467468 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30467468 Supplier Toronto Research Chemicals Supplier No. TRCM31523010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl D-(+)-Lactate
CAS 17392-83-5
Molecular Formula C4H8O3
Quantity 10 g
Synonym (2R)-2-Hydroxypropanoic Acid Methyl Ester, (+)-2-Hydroxypropionic Acid Methyl Ester, (+)-Lactic Acid Methyl Ester, (+)-Methyl 2-Hydroxypropionate, (+)-Methyl Lactate, (R)-Methyl Lactate, D-Methyl Lactate, Methyl (2R)-2-Hydroxypropanoate, Methyl (R)-(+)-Lactate, Methyl (R)-(+)-Lactate, Methyl (R)-2-Hydroxypropanoate, Methyl (R)-2-Hydroxypropionate, Methyl (R)-Lactate, Methyl D-Lactate
InChI Formula InChI=1S/C4H8O3/c1-3(5)4(6)7-2/h3,5H,1-2H3/t3-/m1/s1
SMILES COC(=O)[C@@H](C)O
IUPAC Name methyl (2R)-2-hydroxypropanoate
Molecular Weight (g/mol) 104.1
Formula Weight 104.0473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.