missing translation for 'onlineSavingsMsg'
Learn More

Methyl Desoxycholate-d5, TRC

Product Code. 30465649
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30465649 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30465649 Supplier Toronto Research Chemicals Supplier No. TRCM29823750MG
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spezifikation

Chemical Name or Material Methyl Desoxycholate-d5
CAS 52840-15-0
Molecular Formula C25H37D5O4
Quantity 50 mg
Synonym (3α,5β,12α)-3,12-Dihydroxy-cholan-2,2,3,4,4-d5-24-oic Acid Methyl Ester, Deoxycholic-d5 Acid Methyl Ester, Methyl 7-Deoxycholate-d5
InChI Formula InChI=1S/C25H42O4/c1-15(5-10-23(28)29-4)19-8-9-20-18-7-6-16-13-17(26)11-12-24(16,2)21(18)14-22(27)25(19,20)3/h15-22,26-27H,5-14H2,1-4H3/t15-,16-,17-,18?,19-,20+,21?,22+,24+,25-/m1/s1/i11D2,13D2,17D
SMILES O[C@]1([2H])C([2H])([2H])C[C@@]2(C)[C@@](CCC3C2C[C@H](O)[C@@]4(C)[C@@]3([H])CC[C@@H]4[C@@H](CCC(OC)=O)C)([H])C1([2H])[2H]
Molecular Weight (g/mol) 411.63
Formula Weight 411.3397
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.