missing translation for 'onlineSavingsMsg'
Learn More

Methyl diphenylphosphinite, 99%, ACROS Organics™

Product Code. 10468900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10468900 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10468900 Supplier Thermo Scientific Acros Supplier No. 165290050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 4020-99-9
Color Colorless to Yellow
Density 1.0700g/mL
Boiling Point 149.0°C (6.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C13H13OP
Linear Formula (C6H5)2POCH3
Refractive Index 1.6045 to 1.6065
MDL Number MFCD00048025
Quantity 5 g
Specific Gravity 1.07
Synonym methoxydiphenylphosphine, methyl diphenylphosphinite, diphenylphosphinous acid methyl ester, phosphinous acid, diphenyl-, methyl ester, methoxy diphenyl phosphane, pubchem6470, acmc-209jbw, diphenyl methoxy phosphine
InChI Key OAADXJFIBNEPLY-UHFFFAOYSA-N
SMILES COP(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name methyl diphenylphosphinite
Molecular Weight (g/mol) 216.22
PubChem CID 77636
Formula Weight 216.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl diphenylphosphinite, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl diphenylphosphinite
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.