missing translation for 'onlineSavingsMsg'
Learn More

Methyl Formate, TRC

Product Code. 30473985
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30473985 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473985 Supplier Toronto Research Chemicals Supplier No. TRCM30539010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Formic acid-methyl ester
CAS 107-31-3
Molecular Formula C2 H4 O2
Quantity 10 g
Synonym Formic acid','methyl ester, Methanoic acid methyl ester, Methyl formate, Methyl methanoate, R 611
InChI Formula InChI=1S/C2H4O2/c1-4-2-3/h2H,1H3
SMILES COC=O
IUPAC Name methyl formate
Molecular Weight (g/mol) 60.05
Formula Weight 60.0211
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.