missing translation for 'onlineSavingsMsg'
Learn More

Methyl Homoveratrate, TRC

Product Code. 30480348
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480348 500 mg 500mg
30482144 100 mg 100mg
30465022 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30480348 Supplier Toronto Research Chemicals Supplier No. TRCM323530500MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15964-79-1
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.23
Synonym 2-(3,4-Dimethoxyphenyl)ethanoic Acid Methyl Ester, 3,4-Dimethoxyphenylacetic Acid Methyl Ester, Homoveratric Acid Methyl Ester, Methyl (3,4-Dimethoxyphenyl)acetate, Methyl 2-(3,4-Dimethoxyphenyl)acetate
IUPAC Name methyl 2-(3,4-dimethoxyphenyl)acetate
SMILES COC(=O)Cc1ccc(OC)c(OC)c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H14O4/c1-13-9-5-4-8(6-10(9)14-2)7-11(12)15-3/h4-6H,7H2,1-3H3
Formula Weight 210.0892
Chemical Name or Material Methyl Homoveratrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.