missing translation for 'onlineSavingsMsg'
Learn More

Methyl Hydrazine Sulfate, TRC

Product Code. 30480377
Change view
Click to view available options
Quantity:
500 mg
25 g
100 g
Unit Size:
100g
25g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480377 25 g 25g
30475597 100 g 100g
30468893 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30480377 Supplier Toronto Research Chemicals Supplier No. TRCM31230025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302-15-8
Molecular Formula CH8N2O4S
Molecular Weight (g/mol) 144.15
Synonym MHS, Methylhydrazine Monosulfate, Methylhydrazinium Sulfate, NSC 3802
IUPAC Name methylhydrazine;sulfuric acid
SMILES CNN.OS(=O)(=O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/CH6N2.H2O4S/c1-3-2;1-5(2,3)4/h3H,2H2,1H3;(H2,1,2,3,4)
Formula Weight 144.0205
Chemical Name or Material Methyl Hydrazine Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.