missing translation for 'onlineSavingsMsg'
Learn More

Methyl isobutyrylacetate, 97+%

Product Code. 11362937
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362937 100 g 100g
11352937 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362937 Supplier Thermo Scientific Alfa Aesar Supplier No. L16216.22
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 42558-54-3 | C7H12O3 | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42558-54-3
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00040499
InChI Key HNNFDXWDCFCVDM-UHFFFAOYSA-N
Synonym methyl isobutyrylacetate, methyl 4-methyl-3-oxovalerate, 4-methyl-3-oxovaleric acid methyl ester, iso-butyryl methyl acetate, methyl 4-methyl-3-oxo-pentanoate, methylisobutyrylacetate, acmc-209jq9, ksc237g2d, methyl 4-methyl-3oxopentanoate, methyl4-methyl-3-oxopentanoate
PubChem CID 2759969
IUPAC Name methyl 4-methyl-3-oxopentanoate
SMILES CC(C)C(=O)CC(=O)OC

Specifications

Melting Point -75°C
Density 1.01
Boiling Point 192°C
Flash Point 79°C (174°F)
Refractive Index 1.4265
Quantity 100 g
Beilstein 1857823
Formula Weight 144.17
Percent Purity 98%
Chemical Name or Material Methyl isobutyrylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.