Learn More
Methyl isopropyl carbonate, 97%
Product Code. 11388236
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388236 25 g 25g
11368236 1 g 1g
11378236 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11388236 Supplier Thermo Scientific Alfa Aesar Supplier No. L15314.14
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
CAS: 51729-83-0 | C5H10O3 | 118.13 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51729-83-0
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD01075689
InChI Key RCIJMMSZBQEWKW-UHFFFAOYSA-N
Synonym methyl isopropyl carbonate, isopropyl methyl carbonate, acmc-20akf7, ch3 c o och ch3 2, carbonic acid methylisopropyl ester
PubChem CID 7021530
IUPAC Name methyl propan-2-yl carbonate
SMILES COC(=O)OC(C)C
Specifications
Density 0.975
Boiling Point 116°C to 118°C
Flash Point 27°C (80°F)
Refractive Index 1.383
Quantity 25 g
UN Number UN3272
Beilstein 1744735
Formula Weight 118.13
Percent Purity 97%
Chemical Name or Material Methyl isopropyl carbonate