missing translation for 'onlineSavingsMsg'
Learn More

Methyl L-Tyrosinate, TRC

Product Code. 30471721
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30471721 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30471721

Brand: Toronto Research Chemicals TRCM3325255G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Tyrosine Methyl Ester
CAS 1080-06-4
Molecular Formula C10 H13 N O3
Quantity 5 g
Synonym L-Tyrosine methyl ester, (S)-2-Amino-3-(4-hydroxyphenyl)propionic acid methyl ester, Methyl (2S)-2-amino-3-(4-hydroxyphenyl)propionate, Methyl (2S)-3-(4-hydroxyphenyl)-2-aminopropionate, Methyl (S)-2-amino-3-(4-hydroxyphenyl)propanoate, Methyl L-tyrosinate, Methyl tyrosinate, S-Tyrosine methyl ester, Tyrosine methyl ester
InChI Formula InChI=1S/C10H13NO3/c1-14-10(13)9(11)6-7-2-4-8(12)5-3-7/h2-5,9,12H,6,11H2,1H3/t9-/m0/s1
SMILES COC(=O)[C@@H](N)Cc1ccc(O)cc1
IUPAC Name methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
Molecular Weight (g/mol) 195.22
Formula Weight 195.0895
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.