missing translation for 'onlineSavingsMsg'
Learn More

Methyl Lithocholate, TRC

Product Code. 30464361
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30464361 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464361 Supplier Toronto Research Chemicals Supplier No. TRCM3134205MG
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithocholic Acid Methylester
CAS 1249-75-8
Molecular Formula C25 H42 O3
Quantity 5 mg
Synonym Cholan-24-oic acid','3-hydroxy-','methyl ester','(3α,5β)-, 5β-Cholan-24-oic acid','3α-hydroxy-','methyl ester (8CI), 5β-Cholanic acid','3α-hydroxy-','methyl ester (6CI,7CI), Lithocholic acid methyl ester, Methyl 3α-hydroxy-5β-cholan-24-oate, Methyl 3α-hydroxy-5β-cholanoate, Methyl lithocholate, NSC 63558, NSC 8116
InChI Formula InChI=1S/C25H42O3/c1-16(5-10-23(27)28-4)20-8-9-21-19-7-6-17-15-18(26)11-13-24(17,2)22(19)12-14-25(20,21)3/h16-22,26H,5-15H2,1-4H3/t16-,17-,18-,19+,20-,21+,22+,24+,25-/m1/s1
SMILES COC(=O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name methyl (4R)-4-[(3R,5R,8R,9S,10S,13R,14S,17R)-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
Molecular Weight (g/mol) 390.6
Formula Weight 390.3134
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.