missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl-Lysine Rabbit, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
201.00€ - 437.00€
Specifications
| Antigen | Methyl-Lysine |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Chromatin Immunoprecipitation 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200 |
| Applications | ChIP Assay, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18357672
|
Bio-Techne
NBP3-15955-20UL |
20 μg |
201.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18302352
|
Bio-Techne
NBP3-15955-100UL |
100 μg |
437.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methyl-Lysine Polyclonal antibody specifically detects Methyl-Lysine in All species samples. It is validated for Western Blot, ChIP assay, ImmunoprecipitationSpecifications
| Methyl-Lysine | |
| ChIP Assay, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific | |
| PBS, 50% glycerol, pH7.3 | |
| A synthetic peptide corresponding to a sequence containing Tri-methylated K. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, Chromatin Immunoprecipitation 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| All species | |
| Dimethylysine, Methyl lysine, methyllysine, monomethyl lysine, N epsilon dimethyl lysine, N epsilon mono methyllysine, N epsilon trimethyl lysine, N(epsilon) trimethyl lysine, Trimethyl lysine | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title