missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-toluate, 99%

Product Code. 11436326
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436326 100 g 100g
11446326 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11436326 Supplier Thermo Scientific Alfa Aesar Supplier No. B23111.22
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 99-75-2 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-75-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00008441
InChI Key QSSJZLPUHJDYKF-UHFFFAOYSA-N
Synonym methyl p-toluate, methyl-p-toluate, p-carbomethoxytoluene, methyl 4-toluate, 4-methylbenzoic acid methyl ester, methyl p-methylbenzoate, benzoic acid, 4-methyl-, methyl ester, 4-methoxycarbonyl toluene, p-toluic acid, methyl ester, methyl p-toluenecarboxylate
PubChem CID 7455
IUPAC Name methyl 4-methylbenzoate
SMILES CC1=CC=C(C=C1)C(=O)OC

Specifications

Melting Point 32°C to 35°C
Density 1.06
Boiling Point 221°C to 224°C
Flash Point 89°C (192°F)
Odor Stench
Quantity 100 g
Beilstein 1100609
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material Methyl p-toluate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.