missing translation for 'onlineSavingsMsg'
Learn More

Methyl Propyl Sulfide, TRC

Product Code. 30483254
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483254 500 mg 500mg
30469945 250 mg 250mg
30481405 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30483254 Supplier Toronto Research Chemicals Supplier No. TRCM340913500MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3877-15-4
Molecular Formula C4H10S
Molecular Weight (g/mol) 90.18
IUPAC Name 1-methylsulfanylpropane
SMILES CCCSC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C4H10S/c1-3-4-5-2/h3-4H2,1-2H3
Formula Weight 90.0503
Chemical Name or Material Methyl Propyl Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.