missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red 1 g/L in Water Indicator Solution, Chem Lab

Product Code. 11913343
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11913343 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11913343 Supplier Chem-Lab Supplier No. CL06.1303.0250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Color Red
CAS 845-10-3
Boiling Point 100°C
Melting Point 0°C
Concentration 1 g/L
Density 1 g/mL
Molecular Weight (g/mol) 291.28
Packaging Polyethylene
pH 4.4 to 6.2
Quantity 250 mL
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.