missing translation for 'onlineSavingsMsg'
Learn More

Methyl red sodium salt, For ACS analysis, MP Biomedicals™

Product Code. 11442882 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
11442882 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 11442882

Brand: MP Biomedicals 0215254725

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 845-10-3
Molecular Formula C15H14N3NaO2
Synonym 4-Dimethylaminoazobenzene-2'-carboxylic acid sodium salt
InChI Key GNTPCYMJCJNRQB-UHFFFAOYSA-M
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
IUPAC Name sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
Molecular Weight (g/mol) 291.286
PubChem CID 4465632
Formula Weight 291.3
Color Brown
Quantity 25 g
Physical Form Powder
Chemical Name or Material Methyl red sodium salt, For ACS analysis
Show More Show Less
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.