missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red Sodium Salt, TRC

Product Code. 30467144
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30467144 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30467144

Brand: Toronto Research Chemicals TRCM3328535G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Red Sodium Salt
CAS 845-10-3
Molecular Formula C15H15N3NaO2
Quantity 5 g
Synonym C.I. Acid Red 2, Sodium Salt, Methyl Red Sodium Salt, o-Methyl Red Sodium Salt, 2-[[4-(Dimethylamino)phenyl]azo]-benzoic Acid Sodium Salt
InChI Formula InChI=1S/C15H15N3O2.Na/c1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20;/h3-10H,1-2H3,(H,19,20);/q;+1/p-1
SMILES [Na+].CN(C)c1ccc(cc1)N=Nc2ccccc2C(=O)[O-]
IUPAC Name sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
Molecular Weight (g/mol) 292.29
Formula Weight 291.0984
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.